Learn More
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Melting Point | -73.0°C |
| Color | Colorless to Yellow |
| Density | 1.0900g/mL |
| Boiling Point | 128.0°C to 130.0°C |
| Flash Point | 28°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Linear Formula | ClCH2CH(OCH3)2 |
| Refractive Index | 1.4140 to 1.4160 |
| Mostrar más |
Seguridad y manipulación
- Chloroacetaldehyde dimethyl acetal
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.