missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde dimethyl acetal, 97%

Product Code. 11486017
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486017 500 g 500g
11476017 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486017 Supplier Thermo Scientific Alfa Aesar Supplier No. B25049.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-97-2 | C4H9ClO2 | 124.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-97-2
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00000948
InChI Key CRZJPEIBPQWDGJ-UHFFFAOYSA-N
Synonym chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane
PubChem CID 60986
IUPAC Name 2-chloro-1,1-dimethoxyethane
SMILES COC(CCl)OC

Specifications

Melting Point <-73°C
Density 1.094
Boiling Point 128°C to 130°C
Flash Point 28°C (82°F)
Odor Aromatic
Refractive Index 1.415
Quantity 500 g
UN Number UN1993
Beilstein 1733700
Sensitivity Moisture sensitive
Formula Weight 124.57
Percent Purity 98%
Chemical Name or Material Chloroacetaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.