missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(triphenylphosphine)gold(I), TRC
Specifications
Specifications
| Chemical Name or Material | Chloro(triphenylphosphine)gold(I) |
| CAS | 14243-64-2 |
| Molecular Formula | C18H15AuClP |
| Quantity | 250 mg |
| Synonym | Chloro(triphenylphosphine)gold, (Triphenylphosphine)gold Chloride, (Triphenylphosphine)gold Monochloride, (Triphenylphosphine)gold(1+) Monochloride, (Triphenylphosphine)gold(I) Chloride, AuPPh3Cl, NSC 306388, SKF 40738, Triphenylphosphine Gold Chloride Complex ((C6H5)3P.AuCl), Triphenylphosphineaurous Chloride, Triphenylphosphinechlorogold |
| InChI Formula | InChI=1S/C18H15P.Au.ClH/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h1-15H;;1H/q;+1;/p-1 |
| SMILES | [Cl-].[Au+].c1ccc(cc1)P(c2ccccc2)c3ccccc3 |
| IUPAC Name | gold(1+);triphenylphosphane;chloride |
| Molecular Weight (g/mol) | 494.71 |
| Formula Weight | 494.0265 |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?