missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min

Product Code. 11339407
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339407 5 g 5g
11329407 0.5 g 0.5g
11319407 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339407 Supplier Thermo Scientific Alfa Aesar Supplier No. 041007.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol

Chloro(triphenylphosphine)gold(I) is a common reagent in gold chemistry. It catalyzes certain rearrangement reactions in organic synthesis. It is also used to prepare 1-methylthyminato-N3-triphenylphosphinegold(I) by reacting with 1-methylthymine. It acts as a precursor for the synthesis of gold(I) and gold(III) organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14243-64-2
Molecular Formula C18H15AuClP
Molecular Weight (g/mol) 494.71
MDL Number MFCD00009588
InChI Key IFPWCRBNZXUWGC-UHFFFAOYSA-M
Synonym chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
PubChem CID 10874691
IUPAC Name chlorogold;triphenylphosphane
SMILES Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Assay Percent Range 99.99% (Metal basis, Au 39.3% min.)
Odor Odorless
Quantity 5 g
Solubility Information Soluble in methylene chloride,acetonitrile,benzene and acetone. Insoluble in water and ethanol.
Formula Weight 494.71
Physical Form Powder
Chemical Name or Material Chloro(triphenylphosphine)gold(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.