missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), 99%

Product Code. 10297053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10297053 1 g Glass bottle 1g
10062773 250 mg Glass bottle 250mg
10229623 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10297053 Supplier Thermo Scientific Acros Supplier No. 391610010
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14243-64-2
Molecular Formula C18H15AuClP
Molecular Weight (g/mol) 494.71
MDL Number MFCD00009588
InChI Key IFPWCRBNZXUWGC-UHFFFAOYSA-M
Synonym chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
PubChem CID 10874691
SMILES Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Au)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
IUPAC Name chlorogold;triphenylphosphane
Formula Weight 494.71
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Chloro(triphenylphosphine)gold(I)
Show More Show Less
compliance-icons
Product Identifier
  • Chloro(triphenylphosphine)gold(I)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.