missing translation for 'onlineSavingsMsg'
Learn More

Chloro/Nitrobenzene Mixture 570 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30319997
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319997 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319997 Supplier Dr. Ehrenstorfer Supplier No. DREA50000570ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Chloro-2,4-dinitrobenzene∣1-Chloro-2-nitrobenzene∣1-Chloro-3-nitrobenzene∣1-Chloro-4-nitrobenzene
CAS 97-00-7, 88-73-3, 121-73-3, 100-00-5
Analyte or Component Names 1-Chloro-2,4-dinitrobenzene∣1-Chloro-2-nitrobenzene∣1-Chloro-3-nitrobenzene∣1-Chloro-4-nitrobenzene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1-Chloro-2,4-dinitrobenzene 2000 μg/mL∣1-Chloro-2-nitrobenzene 2000 μg/mL∣1-Chloro-3-nitrobenzene 2000 μg/mL∣1-Chloro-4-nitrobenzene 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.