missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro bis-(4-fluorophenyl)methane, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.2500g/mL |
| Boiling Point | 88°C (0.1 Mbar) |
| Flash Point | 81°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5560 to 1.558 |
| Quantity | 100 g |
| Specific Gravity | 1.25 |
| Show More |
Safety and Handling
Product Identifier
- Chloro bis-(4-fluorophenyl)methane
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?