missing translation for 'onlineSavingsMsg'
Learn More

chloro(1,5-hexadiene)rhodium(I) dimer, Thermo Scientific™

Product Code. 10431723 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10431723 250 mg Glass bottle 250mg
10046172 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431723 Supplier Thermo Scientific Acros Supplier No. 377922500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32965-49-4
Molecular Formula C12H20Cl2Rh2
Molecular Weight (g/mol) 441.02
MDL Number MFCD00064595

Specifications

Melting Point 118.0°C to 120.0°C
Color Golden
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Formula Weight 441.02
Physical Form Crystalline Powder
Chemical Name or Material chloro(1, 5-hexadiene)rhodium(I) dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.