missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)rhodium(I) Dimer, TRC

Product Code. 30366770
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366770 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366770 Supplier Toronto Research Chemicals Supplier No. TRCC2933502G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloro(1,5-cyclooctadiene)rhodium(I) Dimer
CAS 12092-47-6
Molecular Formula C16H24Cl2Rh2
Quantity 2 g
Synonym (1,5-Cyclooctadiene)Rhodium Chloride Dimer, (1,5-Cyclooctadiene) Rhodium(I) Chloride Dimer, (eta4-1,5-Cyclooctadiene)Rhodium Chloride Dimer, Bis((1,5-cyclooctadiene)(Chloro)Rhodium), Bis((μ-chloro)(eta4-1,5-Cyclooctadiene)Rhodium), Bis(1,5-cyclooctadiene)Dirhodium Dichloride, Bis(chloro(1,5-cyclooctadiene)Rhodium), Bis(chloro(eta4-1,5-cyclooctadiene)rhodium), Bis(cyclooctadiene)Dichlorodirhodium, Bis(cyclooctadienerhodium Chloride), Chlororhodium(1,5-cyclooctadiene) Dimer, Cyclooctadiene Rhodium Chloride Dimer, Di-μ-Chlorobis(1,5-cyclooctadiene)Dirhodium, Di-μ-chlorobis(cyclooctadiene) Dirhodium, Di-μ-chlorobis(eta4-1,5-cyclooctadiene)dirhodium, Di-μ-chlorobis(eta4-1,5-cyclooctadiene)Dirhodium(I), Di-μ-chlorobis[(1,2,5,6-eta)-1,5-cyclooctadiene]Dirhodium, Di-μ-chlorobis[(1,5-cyclooctadiene)Rhodium], Di-μ2-chlorobis(1,5-cyclooctadiene)Dirhodium, Dichlorobis(1,5-cyclooctadiene)Dirhodium, Dichlorobis(1,5-cyclooctadiene)Rhodium(I), Dichlorobis(cyclooctadiene)Dirhodium, Rhodium 1,5-Cyclooctadiene chloride dimer, μ-Chloro(1,5-cyclooctadiene)Rhodium(I) Dimer, Bis(1,5-cyclooctadienedirhodium(I) Chloride)
InChI Formula InChI=1S/2C8H12.2ClH.2Rh/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;2*1H;;/q;;;;2*+1/p-2/b2*2-1-,8-7-;;;;
SMILES Cl[Rh].Cl[Rh].C1C\C=C/CC\C=C/1.C2C\C=C/CC\C=C/2
IUPAC Name chlororhodium;(1Z,5Z)-cycloocta-1,5-diene
Molecular Weight (g/mol) 493.08
Formula Weight 491.9365
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.