missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)

Product Code. 15447315
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447315 250 mg 250mg
15437315 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447315 Supplier Thermo Scientific Alfa Aesar Supplier No. 046803.MD
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92390-26-6 | C18H27ClRu | 379.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92390-26-6
Molecular Formula C18H27ClRu
Molecular Weight (g/mol) 379.93
MDL Number MFCD07369035
InChI Key RSZMVRFBKKVJKS-PHFPKPIQSA-M
IUPAC Name Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
SMILES Cl[Ru++].C1C\C=C/CC\C=C/1.Cc1c(C)c(C)c(C)c1C

Specifications

Melting Point 143°C to 147°C
Quantity 250 mg
UN Number UN3395
Synonym Pentamethylcyclopentadienyl(1,5-cyclooctadiene)ruthenium(II) chloride; RuCl(cod)Cp
Formula Weight 379.94
Physical Form Crystalline Solid
Chemical Name or Material Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.