missing translation for 'onlineSavingsMsg'
Learn More

Chlormezanone, MedChemExpress

Product Code. 16438256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438256 Supplier Medchem Express Supplier No. HYB0353100mg
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlormezanone resembles benzodiazepine. The action of Chlormezanone is similar to benzodiazepine-type agents. Chlormezanone is used as an anxiolytic and a muscle relaxant.

Chlormezanone resembles benzodiazepine. The action of Chlormezanone is similar to benzodiazepine-type agents. Chlormezanone is used as an anxiolytic and a muscle relaxant.

Specifications

Chemical Name or Material Chlormezanone
Physical Form Solid
CAS 80-77-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H12ClNO3S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (365.31 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(CC1)N(C)C(C2=CC=C(Cl)C=C2)S1(=O)=O
Molecular Weight (g/mol) 273.74
Formula Weight 273.74
Percent Purity 99.71%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.