missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlormethiazole hydrochloride, Tocris Bioscience™
Potentiates GABAA receptor function
Chemical Identifiers
CAS | 6001-74-7 |
---|---|
Molecular Formula | C6H9Cl2NS |
Molecular Weight (g/mol) | 198.11 |
MDL Number | MFCD00673948 |
InChI Key | OFXYKSLKNMTBHK-UHFFFAOYSA-N |
Synonym | chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1 |
PubChem CID | 19035073 |
IUPAC Name | 5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride |
SMILES | Cl.CC1=C(CCCl)SC=N1 |
Description
Sedative and anticonvulsant which is neuroprotective in a number of animal models. Prevents the degeneration of serotonergic nerve terminals induced by MDMA ('Ecstasy').Chemical Identifiers
6001-74-7 | |
198.11 | |
OFXYKSLKNMTBHK-UHFFFAOYSA-N | |
19035073 | |
Cl.CC1=C(CCCl)SC=N1 |
C6H9Cl2NS | |
MFCD00673948 | |
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1 | |
5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride |
Specifications
6001-74-7 | |
MFCD00673948 | |
OFXYKSLKNMTBHK-UHFFFAOYSA-N | |
5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride | |
19035073 | |
>99% |
C6H9Cl2NS | |
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1 | |
Cl.CC1=C(CCCl)SC=N1 | |
198.11 | |
198.11 | |
Chlormethiazole hydrochloride |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature