missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorisondamine diiodide, Tocris Bioscience™
Nicotinic antagonist; slow offset
Chemical Identifiers
CAS | 96750-66-2 |
---|---|
Molecular Formula | C14H20Cl4I2N2 |
Molecular Weight (g/mol) | 611.937 |
InChI Key | FPNVAOZHQUJJJQ-UHFFFAOYSA-L |
Synonym | chlorisondamine diiodide, 4,5,6,7-tetrachloro-1,3-dihydro-2-methyl-2-2-trimethylammonium ethyl-2h-isoindolium diiodide, chlorisondamine diiodide hplc , white solid, 3-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine diiodide, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-2-trimethylammonio ethyl-2h-isoindolium diiodide, 4,5,6,7-tetrachloro-2-methyl-2-2-trimethylazaniumyl ethyl-2,3-dihydro-1h-isoindol-2-ium diiodide |
PubChem CID | 16078959 |
IUPAC Name | trimethyl-[2-(4,5,6,7-tetrachloro-2-methyl-1,3-dihydroisoindol-2-ium-2-yl)ethyl]azanium;diiodide |
SMILES | C[N+]1(CC2=C(C1)C(=C(C(=C2Cl)Cl)Cl)Cl)CC[N+](C)(C)C.[I-].[I-] |
Description
An exceptionally long lasting nicotinic antagonist (IC50 ∽ 1.6 mM); blockade of central nicotinic responses induced by chlorisondamine can persist for several weeks.Chemical Identifiers
96750-66-2 | |
611.937 | |
chlorisondamine diiodide, 4,5,6,7-tetrachloro-1,3-dihydro-2-methyl-2-2-trimethylammonium ethyl-2h-isoindolium diiodide, chlorisondamine diiodide hplc , white solid, 3-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine diiodide, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-2-trimethylammonio ethyl-2h-isoindolium diiodide, 4,5,6,7-tetrachloro-2-methyl-2-2-trimethylazaniumyl ethyl-2,3-dihydro-1h-isoindol-2-ium diiodide | |
trimethyl-[2-(4,5,6,7-tetrachloro-2-methyl-1,3-dihydroisoindol-2-ium-2-yl)ethyl]azanium;diiodide |
C14H20Cl4I2N2 | |
FPNVAOZHQUJJJQ-UHFFFAOYSA-L | |
16078959 | |
C[N+]1(CC2=C(C1)C(=C(C(=C2Cl)Cl)Cl)Cl)CC[N+](C)(C)C.[I-].[I-] |
Specifications
96750-66-2 | |
chlorisondamine diiodide, 4,5,6,7-tetrachloro-1,3-dihydro-2-methyl-2-2-trimethylammonium ethyl-2h-isoindolium diiodide, chlorisondamine diiodide hplc , white solid, 3-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine diiodide, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-2-trimethylammonio ethyl-2h-isoindolium diiodide, 4,5,6,7-tetrachloro-2-methyl-2-2-trimethylazaniumyl ethyl-2,3-dihydro-1h-isoindol-2-ium diiodide | |
C[N+]1(CC2=C(C1)C(=C(C(=C2Cl)Cl)Cl)Cl)CC[N+](C)(C)C.[I-].[I-] | |
611.937 | |
611.95 | |
Chlorisondamine diiodide |
C14H20Cl4I2N2 | |
FPNVAOZHQUJJJQ-UHFFFAOYSA-L | |
trimethyl-[2-(4,5,6,7-tetrachloro-2-methyl-1,3-dihydroisoindol-2-ium-2-yl)ethyl]azanium;diiodide | |
16078959 | |
>99% |
Safety and Handling
Recommended Storage : Desiccate at +4°C