missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorisondamine Chloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Chlorisondamine Chloride |
| CAS | 69-27-2 |
| Analyte or Component Names | Chlorisondamine Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H20 Cl4 N2 . 2 Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Isoindolium, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-[2-(trimethylammonio)ethyl]-, chloride (1:2), Chlorisondamine Chloride, 1H-Isoindolium, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-[2-(trimethylammonio)ethyl]-, dichloride (9CI), 4,5,6,7-Tetrachloro-2-(2-dimethylaminoethyl)-2-methylisoindolinium chloride methochloride (6CI), Isoindolinium, 4,5,6,7-tetrachloro-2-methyl-2-[2-(trimethylammonio)ethyl]-, dichloride (8CI), Chlorisondamin, Chlorisondamine, Chlorisondamine chloride, Ecolid, Ecolid chloride, Hisindamon, Hisindamone A, N-[(2-Dimethylammonium)ethyl]-4,5,6,7-tetrachloroisoindolinium dimethochloride, SU 3088 |
| InChI Formula | InChI=1S/C14H20Cl4N2.2ClH/c1-19(2,3)5-6-20(4)7-9-10(8-20)12(16)14(18)13(17)11(9)15;;/h5-8H2,1-4H3;2*1H/q+2;;/p-2 |
| Solution Type | Neat |
| SMILES | [Cl-].[Cl-].C[N+](C)(C)CC[N+]1(C)Cc2c(Cl)c(Cl)c(Cl)c(Cl)c2C1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?