missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Chlorine Standard Solution, 25-30 mg/L as Cl2, PourRite Ampules (NIST)

Product Code. 10506225
Click to view available options
Format:
Strip
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10506225

Brand: Hach Lange™ 2630020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For accuracy checks in chlorine determinations.

For accuracy checks in chlorine determinations. Pack of 20 2-mL ampules. Lot-specific concentration displayed on label.

  • Simple Accuracy Checks
  • Minimize waste
  • Reliable results

  • Simple Accuracy Checks
  • Voluette and PourRite Ampule Standards are designed to work directly with Standard Additions accuracy checks in Hach procedures and photometer software. Use the TenSette Pipet to add precise spikes of standard and check percent recovery, to confirm analytical system performance.
  • Minimize waste
  • Open a fresh standard ampule for each accuracy check. Each ampule contains enough standard for one set of standard additions.
  • Reliable results
  • Sealed ampules prevent accidental contamination or evaporation of standard solutions.

    Specifications

  • Concentration: 25 - 30 mg/L Cl₂
  • Description : Chlorine as Cl₂ (NIST)
  • Parameter: Chlorine
  • Platform: Solution
  • Quantity: 20 Ampules - 2 mL

Specifications

Volume (Metric) 20 to 2 mL
Format Strip
Type Solution
Product Identifier
  • Chlorine Standard Solution Ampule 25-30 mg/l as Cl2
Supplemental information
  • MIXTURE LIST-Contains : Hypoclorous acid, potassium hydroxide; caustic potash
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.