Learn More
Lovibond™ Chlorine HR (KI) Tablets
The suitable tablet for the fast result
Brand: Lovibond™ 513000BT
Code : NEW
Additional Details : Weight : 0.30000kg
Description
The suitable tablet for the fast result
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
100 Tablets | |
5 to 200 mg/L, chlorine | |
Chlorine HR (KI) Tablet |