Learn More
Palintest™ Chlorine Dioxide LR Test Kit
Specifications
Specifications
| Concentration | 0 to 2.5 mg/L |
| Quantity | 1 Each |
Safety and Handling
Product Identifier
- REFILL CLO2 LR
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- P264-Wash contaminated skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P405-Store locked up.
- MIXTURE LIST-Contain: lithium hydroxide, boric acid, ammoniumchloride