missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Photometer Tablet Reagents, Chlorine Dioxide and Chlorite (DPD method)

Product Code. 13151461
Change view
Click to view available options
Quantity:
Reagents for 250 tests
Unit Size:
250 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13151461 Reagents for 250 tests 250 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13151461 Supplier Palintest™ Supplier No. AP 052

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Replacement Pack of tablet reagents. Contains reagents for 250 tests.

Chlorine dioxide is used for the disinfection of water in a variety of different applications. Chlorine dioxide is normally generated by reacting chlorine with sodium chlorite solution in specially designed plant and equipment. Water treated with chlorine dioxide may therefore also contain amounts of chlorine and chlorite. For the control of such water treatment systems it is necessary to determine and differentiate between these different residual species.

The Palintest Chlorine Dioxide method provides a precise method of determining chlorine dioxide in treated water. Supplementary procedures provide for the determination of free and combined chlorine and chlorite.

Specifications

Concentration 0 to 10 mg/L ClO2
Description Photometer Tablet Reagents, Replacement Pack
For Use With (Equipment) Palintest™ Photometer 7100, 7500
No. of Tests 250
Quantity Reagents for 250 tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.