missing translation for 'onlineSavingsMsg'
Learn More

Chlorindione, MedChemExpress

Product Code. 16408516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408516 Supplier Medchem Express Supplier No. HYB0918
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorindione (Chlophenadione) is a potent anticoagulant. Chlorindione also is a vitamin K1 antagonist.

Chlorindione (Chlophenadione) is a potent anticoagulant. Chlorindione also is a vitamin K1 antagonist.

Specifications

Chemical Name or Material Chlorindione
CAS 1146-99-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H9ClO2
Quantity 100 mg
Synonym Chlophenadione Indaliton G-25766
Solubility Information DMSO : 100 mg/mL (389.59 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(Cl)C=C2)C(C3=C1C=CC=C3)=O
Molecular Weight (g/mol) 256.68
Formula Weight 256.68
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.