missing translation for 'onlineSavingsMsg'
Learn More

Chlorinated Hydrocarbons Mixture 1011 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30323192
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323192 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323192 Supplier Dr. Ehrenstorfer Supplier No. DREGA09001011DI
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetrachlorobenzene∣1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2-Chloronaphthalene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Hexachloropropene (Perchloropropene)∣Pentachlorobenzene∣Pentachloroethane
CAS 95-94-3, 120-82-1, 95-50-1, 541-73-1, 106-46-7, 91-58-7
Analyte or Component Names 1,2,4,5-Tetrachlorobenzene∣1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2-Chloronaphthalene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Hexachloropropene (Perchloropropene)∣Pentachlorobenzene∣Pentachloroethane
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2,4,5-Tetrachlorobenzene 2000 μg/mL∣1,2,4-Trichlorobenzene 2000 μg/mL∣1,2-Dichlorobenzene 2000 μg/mL∣1,3-Dichlorobenzene 2000 μg/mL∣1,4-Dichlorobenzene 2000 μg/mL∣2-Chloronaphthalene 2000 μg/mL∣Hexachlorobenzene 2000 μg/mL∣Hexachlorobutadiene 2000 μg/mL∣Hexachlorocyclopentadiene 2000 μg/mL∣Hexachloroethane 2000 μg/mL∣Hexachloropropene (Perchloropropene) 2000 μg/mL∣Pentachlorobenzene 2000 μg/mL∣Pentachloroethane 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.