missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chloride Test Kits

Product Code. 11785783
Click to view available options
Type:
Chromate Reagent Test Kit
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11785783

Brand: WTW™ 250353

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Get accurate water test results for waste water, drinking water, ground water, and mineral water/after with chromate reagent test kits.

  • Variety of instrumentation for measuring chloride
  • Chromate reagent test kit
  • For waste water, drinking water, ground water, mineral water/after dilution
  • PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
TRUSTED_SUSTAINABILITY

Specifications

Color Indicator Yellow/Brown
Volume (Metric) 5 mL
For Use With (Application) For waste water, drinking water, ground water, mineral water/after dilution
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
Parameter Specifications Chromate
Range 5 to 125 mg/L
Storage Requirements 15°C to 25°C
Test Method Diphenylcarbazide
Type Chromate Reagent Test Kit
compliance-icons
Product Identifier
  • Chloride Cell Test Method: photometric 5 - 125 mg/l Cl
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : iron(III) nitrate, nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.