missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard for Ion Chromatography, SPEX CertiPrep™

Product Code. 10080522 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10080522 500 mL Triple-leached LDPE bottle 500mL
10570603 125 mL Triple-leached LDPE bottle 125mL
11485918 500 mL Triple-leached LDPE bottle 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10080522 Supplier SPEX Certiprep Supplier No. ASCL92X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cl&superscript_minus;1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
CAS Min % 0.1, 99.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 99.9
Technique Ion Chromatography
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type Certified Reference Material
Solvent or Matrix Water
Quantity 500 mL
Grade IC
Chemical Name or Material Chloride Standard
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.