missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloride Standard for Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£80.00 - £187.00
Chemical Identifiers
CAS | 12125-02-9, 7732-18-5 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
PubChem CID | 25517 |
ChEBI | CHEBI:31206 |
IUPAC Name | amine hydrochloride |
SMILES | N.Cl |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10570603
|
SPEX Certiprep
AS-CL9-2Y |
125 mL | Triple-leached LDPE bottle |
£93.25
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10080522
|
SPEX Certiprep
AS-CL9-2X |
500 mL | Triple-leached LDPE bottle |
£187.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11485918
|
SPEX Certiprep
AS-CL9-2X |
500 mL | Triple-leached LDPE bottle | ||||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cl- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
12125-02-9, 7732-18-5 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
CHEBI:31206 | |
N.Cl |
ClH4N | |
MFCD00011420 | |
25517 | |
amine hydrochloride |
Specifications
12125-02-9, 7732-18-5 | |
0.1, 99.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
ClH4N | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
IC | |
Chloride Standard |
0.1, 99.9 | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011420 | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
H2O |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only