Learn More
Lovibond™ Chloride Combi pack T1 / T2 tablets
Click to view available options
Quantity:
100 Tests
Unit Size:
Pack of 100
Description
Tablet reagent test set
Chloride Combi pack T1 / T2 tablets 100 tests measures 0.5 - 25.0 mg/L.
Specifications
Specifications
| Quantity | 100 Tests |
| Range | Chloride 0.5 to 25.0 mg/L |
| Type | Reagent |
Safety and Handling
Product Identifier
- combi-chloride T1 + T2
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501-Dispose of contents/container to in accordance with all local/ regional/national/international regulations.
- MIXTURE LIST-Contains : sodium hydrogensulphate, potassium monopersulphate triple salt, silver nitrate