Learn More
Palintest™ Chloride (Chloridol) Test Kit
With a range of 0 to 50,000 (NaCl) mg/L, this reliable, well-established, cost-effective test kit is ideal for measuring Chloride (Chloridol).
Brand:  Palintest™ PM268
Code : 04
Additional Details : Weight : 0.80000kg
Product Code. 11984516
  • £111.30 / Pack of 50
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Test kit for measuring Chloride (Chloridol)
  • Range of 0 to 50,000 (NaCl) mg/L
  • X50 Test Chloride 0-50 PPM
Specifications
Specifications
50
0 to 50,000mg/L
Chloride (Chloridol) Test Kit
50 Pack
50 Tests