missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile, Thermo Scientific Chemicals
£25.30 - £650.00
Chemical Identifiers
CAS | 18472-51-0 |
---|---|
Molecular Formula | C34H54Cl2N10O14 |
Molecular Weight (g/mol) | 897.76 |
MDL Number | MFCD00083599 |
InChI Key | YZIYKJHYYHPJIB-UUPCJSQJSA-N |
Synonym | peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate |
PubChem CID | 60148207 |
IUPAC Name | (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane |
SMILES | OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344098
|
Thermo Scientific Alfa Aesar
041385.AC |
25 mL |
£25.30
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334098
|
Thermo Scientific Alfa Aesar
|
100 mL |
£63.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410391
|
Thermo Scientific Alfa Aesar
041385.AP |
500 mL |
£225.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420391
|
Thermo Scientific Alfa Aesar
041385.AY |
4 x 500 mL |
£650.00
2000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorhexidine digluconate, 20% w/v aq. Soln is used as an oral antiseptic agent and anti-microbial agent. It is an active ingredient in germicidal mouthwash, pet shampoo, surgical lubricants and wipes and hand soap. It is utilized in the study to know the role of essential oils for improving skin antisepsis. It plays a vital role as disinfectants and preservative in pharmaceutical products. It is also used as an additive in cosmetics such as creams, toothpaste and deodorants.
Solubility
Miscible with water, ethanol and acetone.
Notes
Store in a cool place. Light sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
18472-51-0 | |
897.76 | |
YZIYKJHYYHPJIB-UUPCJSQJSA-N | |
60148207 | |
OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 |
C34H54Cl2N10O14 | |
MFCD00083599 | |
peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate | |
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane |
Specifications
18472-51-0 | |
1.06 g/mL | |
C34H54Cl2N10O14 | |
Light sensitive | |
YZIYKJHYYHPJIB-UUPCJSQJSA-N | |
(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane | |
60148207 | |
20% w/v aq. soln. | |
Chlorhexidine digluconate, Non-sterile |
Colorless | |
20% w/v aq. soln. | |
MFCD00083599 | |
peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate | |
OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 | |
897.76 | |
897.76 | |
Liquid |
Safety and Handling
P280-P305+P351+P338-P310
H318
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 242-354-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only