missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Diacetate, TRC

Product Code. 30353525
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30353525

Brand: Toronto Research Chemicals TRCC3350551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorhexidine diacetate
CAS 56-95-1
Molecular Formula C22 H30 Cl2 N10 . 2 C2 H4 O2
Quantity 1 g
Synonym 2,4,11,13-Tetraazatetradecanediimidamide, N,N''-bis(4-chlorophenyl)-3,12-diimino-, diacetate (9CI), Biguanide, 1,1'-hexamethylenebis[5-(p-chlorophenyl)-, diacetate (6CI,7CI,8CI), 1,6-Bis(p-chlorophenylbiguanido)hexane diacetate, Arlacide A, Bactigras, Chlorasept 2000, Chlorhexidine acetate, Chlorhexidine diacetate, Chlorzoin, Dosisepsine, EC 40, EC 40 (antibacterial), Hibitane diacetate, Jie-Yin Liquid Disinfectant, NSC 526936
InChI Formula InChI=1S/C22H30Cl2N10.2C2H4O2/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;2*1-2(3)4/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H3,(H,3,4)
SMILES CC(=O)O.CC(=O)O.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1
IUPAC Name acetic acid;1-(4-chlorophenyl)-3-[N-[6-[[N-[N-(4-chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]guanidine
Molecular Weight (g/mol) 625.55
Formula Weight 624.2455
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.