missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhexidine Diacetate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Chlorhexidine diacetate |
| CAS | 56-95-1 |
| Molecular Formula | C22 H30 Cl2 N10 . 2 C2 H4 O2 |
| Quantity | 5 g |
| Synonym | 2,4,11,13-Tetraazatetradecanediimidamide, N,N''-bis(4-chlorophenyl)-3,12-diimino-, diacetate (9CI), Biguanide, 1,1'-hexamethylenebis[5-(p-chlorophenyl)-, diacetate (6CI,7CI,8CI), 1,6-Bis(p-chlorophenylbiguanido)hexane diacetate, Arlacide A, Bactigras, Chlorasept 2000, Chlorhexidine acetate, Chlorhexidine diacetate, Chlorzoin, Dosisepsine, EC 40, EC 40 (antibacterial), Hibitane diacetate, Jie-Yin Liquid Disinfectant, NSC 526936 |
| InChI Formula | InChI=1S/C22H30Cl2N10.2C2H4O2/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;2*1-2(3)4/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H3,(H,3,4) |
| SMILES | CC(=O)O.CC(=O)O.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1 |
| IUPAC Name | acetic acid;1-(4-chlorophenyl)-3-[N-[6-[[N-[N-(4-chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamimidoyl]guanidine |
| Molecular Weight (g/mol) | 625.55 |
| Formula Weight | 624.2455 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction