missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Diacetate, MP Biomedicals™

Product Code. 11468242 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11468242

Brand: MP Biomedicals 0219136180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Effective against a wide range of bacteria, some fungi and some viruses. Chlorhexidine Diacetate, MP Biomedicals is a cationic broad-spectrum antimicrobial agent used as an antiseptic and disinfectant agent. It is effective against both Gram-positive and

Chlorhexidine is a cationic broad-spectrum antimicrobial agent belonging to the bis(biguanide) family. Its mechanism of action involves destabilization of the outer bacterial membrane. It is used primarily as a topical antiseptic/disinfectant in wound healing, at catheterization sites, in various dental applications and in surgical scrubs.

  • It is an agent to prevent gingivitis and was used to study the effects of antimicrobials on dental plaque
  • Commercial ophthalmic products have used this agent to replace thimerosal as a preservative; however, it can cause skin irritation
  • It has been used to study the inactivation of HIV and the immobilization of sperm
  • It was used to study effective skin antisepsis and disinfection of medical devices
TRUSTED_SUSTAINABILITY

Specifications

CAS 56-95-1
Melting Point 151°C to 159°C
Molecular Formula C26H38Cl2N10O4
MDL Number MFCD00150042
Quantity 100 g
Synonym chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane
Solubility Information Soluble in water (0.15g/10mL water with heat), in ethanol (50mg/mL - clear, faint yellow solution), alcohol, glycerol, propylene, glycol, polyethylene glycols.
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1
IUPAC Name acetic acid;(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;hydrate
Molecular Weight (g/mol) 625.56
PubChem CID 43834499
Formula Weight 625.56g/mol
Percent Purity ≥97%
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 0.01% (w/v, 25°C) (Lit.)
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.