missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine diacetate, >97%, MP Biomedicals™

Product Code. 11490432 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11490432

Brand: MP Biomedicals 0219136125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56-95-1
Molecular Formula C26H38Cl2N10O4
Molecular Weight (g/mol) 625.56
MDL Number MFCD00150042
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
Synonym chlorhexidine diacetate, chlorhexidine acetate, bactigras, hibitane diacetate, caswell no. 481e, chlorohexidine diacetate, 10,040 diacetate, chlorhexidine di acetate, chlorhexidine acetate van, unii-5908zuf22y
PubChem CID 43834499
IUPAC Name N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid)
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Quantity 25 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.