missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine, 98%, Thermo Scientific Chemicals

Product Code. 15608715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15608715 1 g 1g
15618715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15608715 Supplier Thermo Scientific Acros Supplier No. 461020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-56-1
Molecular Formula C22H30Cl2N10
Molecular Weight (g/mol) 505.45
InChI Key GHXZTYHSJHQHIJ-UHFFFAOYSA-N
Synonym chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
PubChem CID 9552079
ChEBI CHEBI:3614
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
SMILES C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl

Specifications

Melting Point 134.0°C to 136.0°C
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 505.45
Percent Purity 98%
Chemical Name or Material Chlorhexidine
compliance-icons compliance-icons
Product Identifier
  • Chlorhexidine
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Chlorhexidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.