missing translation for 'onlineSavingsMsg'
Learn More

Chlorflurenol-methyl, TRC

Product Code. 30367390
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367390 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367390 Supplier Toronto Research Chemicals Supplier No. TRCC585278250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorflurenol-methyl ester
CAS 2536-31-4
Molecular Formula C15 H11 Cl O3
Quantity 250 mg
Synonym Fluorene-9-carboxylic acid, 2-chloro-9-hydroxy-, methyl ester (7CI,8CI), 2-Chloro-9-fluorenol-9-carboxylic acid methyl ester, 2-Chloro-9-hydroxy-9H-fluorene-9-carboxylic acid methyl ester, 2-Chloro-9-hydroxyfluorene-9-carboxylic acid methyl ester, CF 125, CME 74050, CME 74050P, Chlorfluorenol IT 3456, Chlorfluorenolmethyl, Chlorflurecol methyl ester, Chlorflurecol-methyl, Chlorflurenol methyl, Chlorflurenol methyl ester, Chloroflurenol methyl ester, EMD 7301, EMD 7301W, EMD-IT 3456, IT 3456, Maintain, Maintain A, Methyl 2-chloro-9-fluorenol-9-carboxylate, Methyl 2-chloro-9-hydroxyfluorene-9-carboxylate, Methyl morphactin, Methyl-2-chloro-9-hydroxyfluorene-9-carboxylic acid, Morphactin IT 3456, TH 417-H
InChI Formula InChI=1S/C15H11ClO3/c1-19-14(17)15(18)12-5-3-2-4-10(12)11-7-6-9(16)8-13(11)15/h2-8,18H,1H3
SMILES COC(=O)C1(O)c2ccccc2c3ccc(Cl)cc13
IUPAC Name methyl 2-chloro-9-hydroxyfluorene-9-carboxylate
Molecular Weight (g/mol) 274.7
Formula Weight 274.0397
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.