missing translation for 'onlineSavingsMsg'
Learn More

Chlorfluazuron, TRC

Product Code. 30133676
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133676 10 g 10g
30040473 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133676 Supplier Toronto Research Chemicals Supplier No. TRCC32920010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71422-67-8
Molecular Formula C20 H9 Cl3 F5 N3 O3
Molecular Weight (g/mol) 540.65
Synonym N-[[[3,5-Dichloro-4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]amino]carbonyl]-2,6-difluorobenzamide, AI 3-29785, Aim, Aim (insecticide), Atabron, CGA 112913, Chlorfluazuron, IKI 7899, Jupiter, Jupiter (pesticide), N-(2,6-Difluorobenzoyl)-N'-[3,5-dichloro-4-(3-chloro-5-trifluoromethyl-2-pyridyloxy)phenyl]urea, PP 145, PP 145 (insecticide), UC 62644
IUPAC Name N-[[3,5-dichloro-4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenyl]carbamoyl]-2,6-difluorobenzamide
SMILES Fc1cccc(F)c1C(=O)NC(=O)Nc2cc(Cl)c(Oc3ncc(cc3Cl)C(F)(F)F)c(Cl)c2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H9Cl3F5N3O3/c21-10-5-9(30-19(33)31-17(32)15-13(24)2-1-3-14(15)25)6-11(22)16(10)34-18-12(23)4-8(7-29-18)20(26,27)28/h1-7H,(H2,30,31,32,33)
Formula Weight 538.963
Chemical Name or Material Chlorfluazuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.