missing translation for 'onlineSavingsMsg'
Learn More

Chlorendic Anhydride, TRC

Product Code. 30365038
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30365038 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365038 Supplier Toronto Research Chemicals Supplier No. TRCC428945500G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorendic Anhydride
CAS 115-27-5
Molecular Formula C9H2Cl6O3
Quantity 500 g
Synonym 4,5,6,7,8,8-Hexachloro-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, 1,4,5,6,7,7-Hexachloro-5-bicyclo[2.2.1]heptene-2,3-dicarboxylic Anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic Anhydride, 1,4,5,6,7,7-Hexachlorobicyclo[2.2.1]-5-heptene-2,3-dicarboxylic Acid Anhydride, 1,4,5,6,7,7-Hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride, 2,3-Dicarboxy-1,4,5,6,7,7-hexachlorobicyclo[2.2.1]hept-5-ene Anhydride, 3,4,5,6,7,7-Hexachloro-1,2,3,6-tetrahydro-3,6-endo-methylenephthalic Anhydride, Chloran 542, Chlorendic Anhydride, HET Anhydride, Hexachloro-5-norbornene-2,3-dicarboxylic Anhydride, Hexachloroendomethylene Tetrahydrophthalic Anhydride, Kayahard CLA, NSC 22229, endo-1,4,5,6,7,7-Hexachloro-1,2,3,4-tetrahydro-5-norbornene-2,3-dicarboxylic Acid Anhydride
InChI Formula InChI=1S/C9H2Cl6O3/c10-3-4(11)8(13)2-1(5(16)18-6(2)17)7(3,12)9(8,14)15/h1-2H
SMILES ClC1=C(Cl)C2(Cl)C3C(C(=O)OC3=O)C1(Cl)C2(Cl)Cl
IUPAC Name 1,7,8,9,10,10-hexachloro-4-oxatricyclo[5.2.1.02,6]dec-8-ene-3,5-dione
Molecular Weight (g/mol) 370.83
Formula Weight 367.8135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.