missing translation for 'onlineSavingsMsg'
Learn More

Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid

Product Code. 11391867
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391867 1000 g 1kg
11371867 50 g 50g
11381867 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391867 Supplier Thermo Scientific Alfa Aesar Supplier No. L07349.0B
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115-27-5 | C9H2Cl6O3 | 370.812 g/mol

Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-27-5
Molecular Formula C9H2Cl6O3
Molecular Weight (g/mol) 370.812
MDL Number MFCD00080438
InChI Key FLBJFXNAEMSXGL-UHFFFAOYSA-N
Synonym chlorendic anhydride, het anhydride, chloran 542, 4,7-methanoisobenzofuran-1,3-dione, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro, 1,4,5,6,7,7-hexachloro-5-norbornene-2,3-dicarboxylic anhydride, chlorendic anhydride van, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, hexachloroendomethylene tetrahydrophthalic anhydride, hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-hexachloro
PubChem CID 8265
SMILES C12C(C(=O)OC1=O)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 235°C to 239°C
Density 1.73
Quantity 1000 g
Beilstein 92693
Merck Index 14,2086
Solubility Information Insoluble in water.
Formula Weight 370.83
Percent Purity 96%
Assay may cont. up to 3% chlorendic acid
Chemical Name or Material Chlorendic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.