missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorazanil, MedChemExpress
Description
Chlorazanil is a triazine derivative and also a new nonmercurial diuretic agent.
Specifications
Specifications
| Chemical Name or Material | Chlorazanil |
| Physical Form | Solid |
| CAS | 500-42-5 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H8ClN5 |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Solubility Information | DMSO : 110 mg/mL (496.28 mM; Need ultrasonic and warming) |
| SMILES | NC1=NC=NC(NC2=CC=C(Cl)C=C2)=N1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?