missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic acid, 98+%

Product Code. 11468733
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468733 500 g 500g
11448733 25 g 25g
11458733 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468733 Supplier Thermo Scientific Alfa Aesar Supplier No. A10493.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-88-7 | C6H2Cl2O4 | 208.978 g/mol

Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

Solubility
Soluble in hot water and methanol. Insoluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-88-7
Molecular Formula C6H2Cl2O4
Molecular Weight (g/mol) 208.978
MDL Number MFCD00001596
InChI Key IPPWILKGXFOXHO-UHFFFAOYSA-N
Synonym chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy
PubChem CID 66604
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O

Specifications

Melting Point >300°C (decomposition)
Quantity 500 g
Beilstein 1875040
Merck Index 14,2079
Solubility Information Soluble in hot water and methanol. Insoluble in organic solvents.
Formula Weight 208.99
Percent Purity ≥98%
Chemical Name or Material Chloranilic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.