missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, TRC

Product Code. 30041091
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041091 50 mg 50mg
30040910 1 g 1g
30005715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041091 Supplier Toronto Research Chemicals Supplier No. TRCC32503050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11 H12 Cl2 N2 O5
Molecular Weight (g/mol) 323.13
Synonym Acetamide, 2,2-dichloro-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-, [R-(R*,R*)]-, Acetamide, 2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]-, D-threo-(-)- (8CI), Levomycetin (7CI), 2,2-Dichloro-N-[(1R,2R)-2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]acetamide, (-)-Chloramphenicol, Ak-Chlor, Alficetyn, Amphenicol, Amphicol, Amseclor, Anacetin, Aquamycetin, Austracol, CAF, CAF (pharmaceutical), CAM, CAP, CPh, Catilan, Chemicetin, Chemicetina, Chlomycol, Chloramex, Chloramphenicol, Chloramsaar, Chlorasol, Chlorcetin, Chloricol, Chlorocaps, Chlorocid, Chlorocide, Chlorocidin C, Chlorocidin C tetran, Chlorocin, Chloromycetin, Chloronitrin, Chloroptic, Ciplamycetin, Cloramfen, Cloramficin, Cloramicol, Clorocyn, Cloromisan, Cylphenicol, D-(-)-Chloramphenicol, D-(-)-threo-1-(4-Nitrophenyl)-2-dichloroacetamido-1,3-propanediol, D-(-)-threo-1-p-Nitrophenyl-2-dichloracetamido-1,3-propanediol, D-(-)-threo-1-p-Nitrophenyl-2-dichloroacetylamino-1,3-propanediol, D-(-)-threo-2-Dichloroacetamido-1-p-nitrophenyl-1,3-propanediol, D-(-)-threo-Chloramphenicol, D-Chloramphenicol, D-threo-(1R,2R)-1-p-Nitrophenyl-2-dichloroacetamido-1,3-propanediol, D-threo-Chloramphenicol, D-threo-N-Dichloroacetyl-1-p-nitrophenyl-2-amino-1,3-propanediol, Detreomycin, Embacetin, Enicol, Enteromycetin, Farmicetina, Fenicol, Globenicol, Gloveticol, Halcetin, I 337A, Intramycetin, Ismicetina, Juvamycetin, Kamaver, Kemicetine, Klorita, Klorocid S, Leukomyan, Leukomycin, Levocin, Levomicetina, Levomitsetin, Levoplast, Levosin, Levovetin, Loromisin, Mastiphen, Micloretin, Micoclorina, Microcetina, Mychel, Mychel-Vet, Mycinol, Myclocin, Mycochlorin, NSC 3069, Normimycin V, Novomycetin, Ocuphenicol, Opclor, Ophthochlor, Pantovernil, Paraxin, Quemicetina, Ronphenil, Septicol, Sintomicetin, Sintomicetina, Sintomicetine R, Sno Phenicol, Soluthor, Stanomycetin, Synthomycetin, Tevcocin, Tifomycine, Treomicetina, Unimycetin, Veticol, Viceton
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O)c1ccc(cc1)[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)/t8-,9-/m1/s1
Formula Weight 322.0123
Percent Purity >95%
Chemical Name or Material Chloramphenicol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.