missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, Mikromol™

Product Code. 18053580
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
18053580 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 18053580

Brand: Mikromol MM0516.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Chloramphenicol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloramphenicol
CAS 56-75-7
Analyte or Component Names Chloramphenicol
Shipping Condition Room Temperature
Molecular Formula C11 H12 Cl2 N2 O5
Certifications/Compliance ISO 17025
Synonym Chloramphenicol, Acetamide, 2,2-dichloro-N-[(1R,2R)-2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-, Acetamide, 2,2-dichloro-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-, [R-(R*,R*)]-, Levomycetin (7CI), (-)-Chloramphenicol, Ak-Chlor, Alficetyn, Amphenicol, Amphicol, Amseclor, Anacetin, Aquamycetin, Austracol, CAF, CAF (pharmaceutical), CAM, CAP, CPh, Catilan, Chemicetin, Chemicetina, Chlomycol, Chloramex, Chloramsaar, Chlorasol, Chlorcetin, Chloricol, Chlorocaps, Chlorocid, Chlorocide, Chlorocidin C, Chlorocidin C tetran, Chlorocin, Chloromycetin, Chloronitrin, Chloroptic, Ciplamycetin, Cloramfen, Cloramficin, Cloramicol, Clorocyn, Cloromisan, Cylphenicol, D-(-)-Chloramphenicol, D-(-)-threo-1-(4-Nitrophenyl)-2-dichloroacetamido-1,3-propanediol, D-(-)-threo-1-p-Nitrophenyl-2-dichloracetamido-1,3-propanediol, D-(-)-threo-1-p-Nitrophenyl-2-dichloroacetylamino-1,3-propanediol, D-(-)-threo-2-Dichloroacetamido-1-p-nitrophenyl-1,3-propanediol, D-(-)-threo-Chloramphenicol, D-Chloramphenicol, D-threo-(1R,2R)-1-p-Nitrophenyl-2-dichloroacetamido-1,3-propanediol, D-threo-Chloramphenicol, D-threo-N-Dichloroacetyl-1-p-nitrophenyl-2-amino-1,3-propanediol, Detreomycin, Embacetin, Enicol, Enteromycetin, Farmicetina, Fenicol, Globenicol, Gloveticol, Halcetin, I 337A, Intramycetin, Ismicetina, Juvamycetin, Kamaver, Kemicetine, Klorita, Klorocid S, Leukomyan, Leukomycin, Levocin, Levomicetina, Levomitsetin, Levoplast, Levosin, Levovetin, Loromisin, Mastiphen, Micloretin, Micoclorina, Microcetina, Mychel, Mychel-Vet, Mycinol, Myclocin, Mycochlorin, NSC 3069, Normimycin V, Novomycetin, Ocuphenicol, Opclor, Ophthochlor, Pantovernil, Paraxin, Quemicetina, Ronphenil, Septicol, Sintomicetin, Sintomicetina, Sintomicetine R, Sno Phenicol, Soluthor, Stanomycetin, Synthomycetin, Tevcocin, Tifomycine, Treomicetina, Unimycetin, Veticol, Viceton, Acetamide, 2,2-dichloro-N-[beta-hydroxy-alpha-(hydroxymethyl)-p-nitrophenethyl]-, D-threo-(-)- (8CI)
InChI Formula InChI=1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)/t8-,9-/m1/s1
SMILES OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O)c1ccc(cc1)[N+](=O)[O-]
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Molecular Weight (g/mol) 323.13
Quantity 500 mg
Formula Weight 322.0123 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.