missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, MedChemExpress

Product Code. 16480838
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16480838 5 g 5g
16432109 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16480838

Brand: Medchem Express HYB0239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chloramphenicol is an orally active, potent and broad-spectrum antibiotic.

Chloramphenicol shows antibacterial activity. Chloramphenicol represses the oxygen-labile transcription factor and hypoxia inducible factor-1 alpha (HIF-1α) in hypoxic A549 and H1299 cells. Chloramphenicol suppresses the mRNA levels of vascular endothelial growth factor (VEGF) and glucose transporter 1, eventually decreasing VEGF release. Chloramphenicol can be used for anaerobic infections and lung cancer research.

Specifications

Chemical Name or Material Chloramphenicol
Physical Form Solid
CAS 56-75-7
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C11H12Cl2N2O5
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 150 mg/mL (464.21 mM) ∣H2O : 3.06 mg/mL (9.47 mM; Need ultrasonic)
SMILES O=C(N[C@H](CO)[C@H](O)C1=CC=C([N+]([O-])=O)C=C1)C(Cl)Cl
Molecular Weight (g/mol) 323.13
Formula Weight 323.13
Percent Purity 98.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.