missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramphenicol Crystalline MP Biomedicals
Click to view available options
Quantity:
5 g
25 g
500 g
Specifications
Density | 1.586 g/mL |
Quantity | 5 g |
Formula Weight | 323.15 |
Physical Form | Crystalline |
Chemical Name or Material | Chloramphenicol |
Safety and Handling
Product Identifier
- Chloramphenicol
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local, regional, national and international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'