missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramphenicol Acetyltransferase Antibody (CAT-1), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50129V
This item is not returnable.
View return policy
Description
Chloramphenicol Acetyltransferase Monoclonal specifically detects Chloramphenicol Acetyltransferase in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Chloramphenicol Acetyltransferase | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| CAT-1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CAT, CAT II, CATB, CatD, Chloramphenicol acetyltransferase 2, Chloramphenicol acetyltransferase II, cmlA | |
| Full length Chloramphenicol Acetyltransferase | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction