missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, 99.78%, MP Biomedicals™

Product Code. 11441861 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441861 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441861 Supplier MP Biomedicals Supplier No. 0219032180
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56-75-7
Color White to Cream
Density 1.586g/mL
Assay Percent Range 0.9978
Molecular Formula C11H12Cl2N2O5
Quantity 100 g
Synonym chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Molecular Weight (g/mol) 323.126
PubChem CID 5959
ChEBI CHEBI:17698
Formula Weight 323.15
Physical Form Crystalline Powder
Chemical Name or Material Chloramphenicol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.