missing translation for 'onlineSavingsMsg'
Learn More

Melford Chloramphenicol

Product Code. 30018189
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
30018189 25 g 25g
30018188 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 30018189

Brand: Melford C6100025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloramphenicol is an antibiotic that belongs to the class of antibiotics known as the phenicols. Chloramphenicol is effective against a broad range of Gram-positive and Gram-negative bacteria.

In the field of microbiology and cell culture, Chloramphenicol is commonly used as a selective agent. It inhibits bacterial protein synthesis by targeting the 50S ribosomal subunit, preventing the growth of bacteria.

Researchers add Chloramphenicol to growth media or solutions to selectively allow the growth of cells or organisms that have been genetically modified to carry a resistance gene for Chloramphenicol. This gene allows the cells to survive and propagate in the presence of the antibiotic, while non-transformed cells are inhibited.

Chloramphenicol also finds applications in molecular biology and genetic engineering research. It can be used to maintain and propagate plasmids or other vectors that contain the Chloramphenicol resistance gene.

Additionally, it is employed in studies involving gene expression and protein synthesis.

Specifications

Chemical Name or Material Chloramphenicol
CAS 56-75-7
Quantity 25 g
Molecular Formula C11H12Cl2N2O5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.