missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T Trihydrate, TRC

Product Code. 30132552
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132552 100 g 100g
30135808 10 g 10g
17952925 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132552 Supplier Toronto Research Chemicals Supplier No. TRCC315160100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7080-50-4
Molecular Formula 3C7H7ClNO2S.3Na.3H2O
Molecular Weight (g/mol) 736.98
Synonym Tosylchloramide sodium, Sodium N-chloro-4-methylbenzene-sulfonimidate trihydrate, Benzenesulfonamide, N-chloro-4-methyl-, sodium salt, hydrate (1:1:3), Benzenesulfonamide, N-chloro-4-methyl-, sodium salt, trihydrate (9CI), Sodium, (N-chloro-p-toluenesulfonamido)-, trihydrate (8CI), Chloramine-T trihydrate, Tosylchloramide sodium trihydrate
IUPAC Name sodium;chloro(p-tolylsulfonyl)azanide;hydrate
SMILES O.O.O.[Na+].[Na+].[Na+].Cc1ccc(cc1)S(=O)(=O)[N-]Cl.Cc2ccc(cc2)S(=O)(=O)[N-]Cl.Cc3ccc(cc3)S(=O)(=O)[N-]Cl

Specifications

Quantity 100 g
InChI Formula InChI=1S/3C7H7ClNO2S.3Na.3H2O/c3*1-6-2-4-7(5-3-6)12(10,11)9-8;;;;;;/h3*2-5H,1H3;;;;3*1H2/q3*-1;3*+1;;;
Formula Weight 734.9668
Chemical Name or Material Tosylchloramide Sodium Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.