missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, ACS, 98.0-103.0%

Product Code. 11304498
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304498 500 g 500g
11394488 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304498 Supplier Thermo Scientific Alfa Aesar Supplier No. 042374.36
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

Reagent for selective oxidation of methionine | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

Reagent for selective oxidation of methionineChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
Synonym benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
PubChem CID 122197679
IUPAC Name N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Melting Point 167°C to 170°C
Color White to Yellow
pH 8 to 10
Physical Form Crystalline
Quantity 500 g
UN Number UN3263
Beilstein 3924168
Flash Point 192°C (377°F)
Merck Index 14,2075
Solubility Information Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Formula Weight 281.69 (227.67 Anhydrous)
Percent Purity 98.0 to 103.0%
Grade ACS
Odor Chlorine-like
Sensitivity Moisture sensitive
Chemical Name or Material Chloramine-T trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.