Learn More
Chloramine-T trihydrate, ACS, 98.0-103.0%
Description
Reagent for selective oxidation of methionineChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 167°C to 170°C |
| Physical Form | Crystalline |
| Quantity | 100 g |
| UN Number | UN3263 |
| Beilstein | 3924168 |
| Flash Point | 192°C (377°F) |
| Merck Index | 14,2075 |
| Solubility Information | Soluble in water and ethyl alcohol. Insoluble in benzene and ethers. |
| Formula Weight | 281.69 (227.67 Anhydrous) |
| Percent Purity | 98.0 to 103.0% |
| Show More |
Safety and Handling
- Chloramine-T trihydrate
- Danger
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- EUH031-Contact with acids liberates toxic gas.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Chloramine-T trihydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.