missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T trihydrate, 98%

Product Code. 11499912
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11499912

Brand: Thermo Scientific Alfa Aesar A12044.0I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol

Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

Solubility
Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.

Notes
Incompatible with strong oxidizing agents, acids and ammonia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
Synonym benzenesulfonamide, n-chloro-4-methyl-, sodium salt, trihydrate
PubChem CID 122197679
IUPAC Name N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Melting Point 167°C to 170°C
pH 8 to 10
Quantity 5000 g
Assay Percent Range 98%
UN Number UN3263
Beilstein 3924168
Flash Point 192°C (377°F)
Merck Index 14,2075
Solubility Information Soluble in water and ethyl alcohol. Insoluble in benzene and ethers.
Formula Weight 281.69 (227.67 Anhydrous)
Percent Purity 98%
Odor Chlorine-like
Sensitivity Moisture sensitive
Chemical Name or Material Chloramine-T trihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloramine-T trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Chloramine-T trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.