missing translation for 'onlineSavingsMsg'
Learn More

Chloramine-T (sodium salt) trihydrate, BAKER ANALYZED™, J.T. Baker™

Product Code. 10061130 Shop All J.T.Baker Products
Change view
Click to view available options
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10061130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10061130 Supplier JT Baker Supplier No. 1255.0250
  • ONLINE EXCLUSIVE PRICE
    £117.00
    £95.52 / 250g
    Save £21.48
    18% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point 167°C to 170°C
CAS 7080-50-4
Quantity 250 g
Assay Percent Range min. 24.5%
Molecular Formula C7H13ClNNaO5S
MDL Number MFCD00149066
Flash Point 92°C
Synonym Chloramine T sodium salt trihydrate , Tosylchloramide sodium trihydrate
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
IUPAC Name sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
Molecular Weight (g/mol) 281.68
Packaging Plastic bottle for solids
Density 1.43 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.