missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Description
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP
Specifications
Melting Point | 170°C |
Color | White |
pH | 8 to 10 |
Physical Form | Powder/Solid |
Quantity | 250 g |
Formula Weight | 281.69g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'